Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120275 Supplier Invitrogen™ Supplier No. PA5120275
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The target is usually found in the following locations: Nucleus. Immunogen sequence: QMIKENQEKT EDQYEENSHL TSVPHWINSN TLGFGTEQAP YGMQTQNYPK GGLLDSMCPA STPSVLSSEQ EFQMFPKSRL SSVSVTYCSV SQDFPGSNLN LLTNNSGTPK DHDSPENGAD SFESSDSLLQ SWNSQSSLLD VQRVPSFESF EDDCSQSLCL NKPTMSFKDY IQERSDPVEQ G.

ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ETS2
Gene Accession No. 0
Gene Alias AU022856; avian erythroblastosis virus E26 (v-ets) oncogene homolog 2; E26 avian leukemia oncogene 2, 3' domain; ETS proto-oncogene 2, transcription factor; Ets2; Ets-2; ETS2IT1; oncogene ETS-2; protein C-ets-2; v-ets avian erythroblastosis virus E2 oncogene homolog 2; v-ets avian erythroblastosis virus E26 oncogene homolog 2; v-ets erythroblastosis virus E26 oncogene homolog 2
Gene Symbols ETS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-345 of human ETS2 (NP_005230.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304063
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.