Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ets21C Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144544 Supplier Invitrogen™ Supplier No. PA5144544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ets21C Polyclonal Antibody for Western Blot, ELISA

ETS21C is a gene that is found in the fruit fly Drosophila melanogaster. It belongs to the ETS transcription factor family, which is a group of proteins that play important roles in regulating gene expression. ETS21C is specifically involved in various developmental processes in Drosophila, including embryonic development, cell differentiation, and tissue patterning. The specific functions and mechanisms of ETS21C are still being studied, and further research is necessary to fully understand its role in cellular processes and its potential implications for Drosophila development and physiology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ets21C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.623 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Ets21C
Gene Accession No. P29776
Gene Alias ETS-6
Gene Symbols Ets21C
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) Ets21C protein (165-334aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 33229
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.