Learn More
Cytiva Ettan IPGphor Manifold Light
GSA_VA
Catalog No. 45002361 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF Unit
Catalog No. For Use With (Equipment)
45-002-361 Ettan IPGphor 3 IEF Unit
1 options
Catalog No. 45-002-361 Supplier Cytiva Supplier No. 11002688
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
For use with Ettan IPGphor 3 isoelectric focusing unit.
  • Cup loading manifold light
  • With plastic tray
  • For routine general screening applications
  • Use sample cups to increase protein loads
  • Made from leading-edge polymeric material
  • Easy to handle and robust
  • Speeds up processing of micropreparative loads in first-dimension analysis
  • Can handle 12 IPG strips simultaneously
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Highly suitable for routine general screening applications
For Use With (Equipment) Ettan IPGphor 3 IEF Unit
Type Ettan IPGphor Cup Loading Manifold Light with Plastic Tray
Includes 1 x Cleaning brush, 2 x Electrode assembly, 1 x Spirit level, 20 x Sample cup, 1 x Sample cup insertion tool, 60 x Paper electrode wicks, 60 x Paper bridge pads, 1 x Manifold, 1 x Forceps, 1 x 60 mL cleaning solution and, User Manual
Material Leading-edge Polymeric Material
No. of Gels 12
Quantity Complete packsize