Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV1 Monoclonal Antibody (1C8B6)
GREENER_CHOICE

Catalog No. PIMA515461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515461 Supplier Invitrogen™ Supplier No. MA515461
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15461 targets ETV1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15461 immunogen is purified recombinant fragment of ETV1 (aa1-191) expressed in E. Coli..

ETV1: ets variant gene 1. Several members of the Ets gene family encode sequence-specific DNA binding proteins that recognize DNA sequences with a centrally located 5'-GGAA-3' element. All of the Ets proteins recognize the same central core sequence but each protein interacts with unique sequences that flank this core. ER81 (also designated ETV1) binds the motif 5'-CGGAA/T-3'. ER81 is highly expressed in brain, testis, lung and heart. ER81 is also moderately expressed in spleen, pancreas, colon and small intestine. During development, ER81 displays a unique expression patterns which suggests that this transcriptional factor might play an important role in organogenesis. ERK-1 activates ER81 transcriptional activity while MAPKAP kinase 2 inhibits ER81.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C8B6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Etv1
Gene Accession No. P50549
Gene Alias DKFZp781L0674; Er81; ets related protein 81; ETS transcription factor Er81; ETS translocation variant 1; ets variant 1; ets variant gene 1; ETS variant transcription factor 1; ets-related protein 81; Etsrp81; Etv1; MGC104699; MGC120533; MGC120534; MGC137631 protein
Gene Symbols Etv1
Host Species Mouse
Immunogen Purified recombinant fragment of ETV1 (aa1-191) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.