Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETV1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen ETV1
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125861
SDP
View Documents
Novus Biologicals
NBP310480100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ETV1 Polyclonal specifically detects ETV1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ETV1
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Stem Cell Markers
PBS buffer, 2% sucrose
2115
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Polyclonal
Purified
RUO
Human
DKFZp781L0674, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534
The immunogen is a synthetic peptide directed towards the middle region of human ETV1 (NP_004947). Peptide sequence PDNQRPLLKTDMERHINEEDTVPLSHFDESMAYMPEGGCCNPHPYNEGYV
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.