Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ETV2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542249
Description
Peptide sequence: DTPTATAETC WKGTSSSLAS FPQLDWGSAL LHPEVPWGAE PDSQALPWSG Sequence homology: Human: 100%.
Binds to DNA sequences containing the consensus pentanucleotide 5'-CGGA[AT]-3'.
Specifications
| ETV2 | |
| Polyclonal | |
| Unconjugated | |
| ETV2 | |
| ER71; ets related protein 71; ETS translocation variant 2; ETS variant 2; ets variant gene 2; ETS variant transcription factor 2; ets-related protein 71; Etsrp71; ETV2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2116 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00321 | |
| ETV2 | |
| Synthetic peptide directed towards the N-terminal of human ETV2 (aa 40-89). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction