Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568762 Supplier Invitrogen™ Supplier No. PA568762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Mouse: 100%; Rat: 92%.

Binds to DNA sequences containing the consensus pentanucleotide 5'-CGGA[AT]-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ETV2
Gene Accession No. P41163
Gene Alias ER71; ets related protein 71; ETS translocation variant 2; ETS variant 2; ets variant gene 2; ETS variant transcription factor 2; ets-related protein 71; Etsrp71; ETV2
Gene Symbols ETV2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse ETV2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14008
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.