Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552107 Supplier Invitrogen™ Supplier No. PA552107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKEEHTQEEG TVPSRTIEEE KGTIFARPAA PPIWPSVPIS TPSGEPLEVT EDSEDRPGKE PSAPEKKEDA LMPPKLRLKR RWNDDPEARE LSKSGKFLWN G Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.

Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV3
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ETV3
Gene Accession No. P41162
Gene Alias AI414410; bA110J1.4; ETS domain transcriptional repressor PE1; ETS translocation variant 3; ETS variant 3; ets variant gene 3; ets variant gene 3, ETS family transcriptional repressor; ETS-domain protein; ETS-domain transcriptional repressor; Etv3; Mets; Mitogenic Ets transcriptional suppressor; mitogenic Ets transcriptional suppressor METS; Pe1; PE-1; SH2 domain containing phosphatase anchor protein 1; Spap1
Gene Symbols ETV3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ETV3. Recombinant protein control fragment (Product #RP-102505).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.