Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Etv3l Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569504
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Yeast: 91%; Zebrafish: 100%.
Transcriptional regulator.
Specifications
Etv3l | |
Polyclonal | |
Unconjugated | |
ETV3L | |
ETS translocation variant 3-like protein; ETS variant 3 like; ets variant 3-like; ets variant gene 3-like; ETS variant transcription factor 3 like; ETV3L; RGD1562620 | |
Rabbit | |
Affinity chromatography | |
RUO | |
499651 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
ETV3L | |
synthetic peptide directed towards the middle region of rat Etv3l. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction