Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETV3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569478 Supplier Thermo Scientific Supplier No. PA569478
Add to Cart
Add to Cart

ETV3L Polyclonal Antibody

ETV3L Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV3L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ETV3L
Gene Accession No. Q6ZN32
Gene Alias ETS translocation variant 3-like protein; ETS variant 3 like; ets variant 3-like; ets variant gene 3-like; ETS variant transcription factor 3 like; ETV3L; RGD1562620
Gene Symbols ETV3L
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ETV3L.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440695
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.