Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589489 Supplier Invitrogen™ Supplier No. PA589489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat lung, Rat heart; Cellular Location: Nucleus Immunogen sequence: PDFHSENLAF HSPTTRIKKE PQSPRTDPAL SCSRKPPLPY HHGEQCLYSS AYDPPRQIAI KSPAPGALGQ SPLQPFPRAE QRNFLRSSGT SQPHPGHGYL.

ETV4: ets variant gene 4 (E1A enhancer binding protein, E1AF), also known as PEA3. Several members of the Ets gene family are known to encode sequencespecific DNA binding proteins. These include mouse PU. 1, mouse and human Ets-1, Drosophila E74, chicken and human Ets-2 and rat GABP-a. Each of these proteins recognizes similar motifs in DNA that share a centrally located 5'-GGAA-3' element. For instance, PEA3 binds the motif 5'-AGGAAG-3' (the PEA-3 motif), but does not bind to the sequence 5'-AGGAAC-3', recognized by PU. 1, although PU. 1 binds equally well to both sequences. It appears that all of the Ets proteins recognize the same central core sequence but that each protein interacts with unique sequences that flank this core. PEA3 is expressed at readily detectable levels in cells of epithelial and fibroblastic origin but is not expressed in hematopoietic cells. This is in contrast to other members of the Ets gene family, such as Ets-1, Ets-2 and Fli-1, each of which is expressed primarily in cells of hematopoietic origin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ETV4
Gene Accession No. P28322
Gene Alias Adenovirus E1A enhancer-binding protein; AW414408; E1A F; E1AF; E1A-F; ETS translocation variant 4; ets variant 4; ets variant gene 4 (E1A enhancer binding protein, E1AF); ets variant gene 4 (E1A enhancer-binding protein, E1AF); ETS variant protein 4; Etv4; EWS protein/E1A enhancer binding protein chimera; PEA3; Pea-3; PEA3 protein; PEAS3; Polyomavirus enhancer activator 3; POLYOMAVIRUS ENHANCER ACTIVATOR 3 (PEA3 PROTEIN) (ETS TRANSLOCATION VARIANT 4); polyomavirus enhancer activator 3 homolog; polyomavirus enhancer activator-3; Protein PEA3
Gene Symbols ETV4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 79-178 of human ETV4 (NP_0019771).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18612, 360635
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.