Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530023 Supplier Invitrogen™ Supplier No. PA530023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: ERM-transfected 293T. Predicted reactivity: Mouse (98%), Rat (95%), Pig (98%), Chicken (88%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ETS variant 5, transcription factor of the ETS family, divergent member of the winged helix-turn-helix superfamily, expressed in brain, placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV5
Applications ChIP Assay, Western Blot, Immunocytochemistry, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene ETV5
Gene Accession No. P41161
Gene Alias 1110005E01Rik; 8430401F14Rik; ERM; ETS translocation variant 5; ets variant 5; ets variant gene 5; ets variant gene 5 (ets-related molecule); ets-related molecule; ets-related protein ERM; ETV5
Gene Symbols ETV5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ERM. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2119
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.