Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ETV5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566555
Description
Immunogen sequence: AAGPVQGVGPA PAPHSLPEPG PQQQTFAVPR PPHQPLQMPK MMPENQYPSE QRFQRQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
ETS variant 5, transcription factor of the ETS family, divergent member of the winged helix-turn-helix superfamily, expressed in brain, placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
Specifications
ETV5 | |
Polyclonal | |
Unconjugated | |
ETV5 | |
1110005E01Rik; 8430401F14Rik; ERM; ETS translocation variant 5; ets variant 5; ets variant gene 5; ets variant gene 5 (ets-related molecule); ets-related molecule; ets-related protein ERM; ETV5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2119 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P41161 | |
ETV5 | |
Recombinant Human ETV5. Recombinant protein control fragment (Product #RP-105253). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction