Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101108 Supplier Invitrogen™ Supplier No. PA5101108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ETV6 / Tel.

This gene encodes an ETS family transcription factor. The product of this gene contains two functional domains: a N-terminal pointed (PNT) domain that is involved in the protein-protein interactions with itself and other proteins, and a C-terminal DNA-binding domain. Gene knockout studies in mice suggest that it is required for hematopoiesis and Maintenance of the developing vascular network. This gene is known to be involved in a large number of chromosomal rearrangements associated with leukemia and congenital fibrosarcoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Etv6
Gene Accession No. P41212, P97360
Gene Alias AW123102; AW557856; CGI-108; CSL4; Csl4p; ETS translocation variant 6; ETS variant 6; ets variant gene 6 (TEL oncogene); ETS-related protein Tel1; Etv6; hCsl4p; p13; RP11-452K12.9; SKI4; Ski4p; Tel; TEL/ABL; Tel1; TEL1 oncogene; THC5; Transcription factor ETV6
Gene Symbols Etv6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ETV6(Accession P41212), corresponding to amino acid residues E231-P281.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14011, 2120, 312777
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.