Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETV6/Tel Antibody (SR1935), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322355100UL
Description
ETV6/Tel Monoclonal antibody specifically detects ETV6/Tel in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
ETV6/Tel | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1935 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
ETS translocation variant 6, ets variant 6, ets variant gene 6 (TEL oncogene), ETS-related protein Tel1, Tel, TEL/ABL, TEL1, TELTEL1 oncogene, transcription factor ETV6 | |
A synthesized peptide derived from human ETV6/Tel (Uniprot #: P41212) | |
100 μg | |
Cancer | |
2120 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction