Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETV7 Antibody (OTI3B2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271938H
Description
ETV7 Monoclonal specifically detects ETV7 in Human samples. It is validated for Western Blot.Specifications
ETV7 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ETV7 | |
Recombinant protein fragment corresponding to amino acids 90-341 of human ETV7 (NP_057219) produced in E.coli. | |
0.1 mL | |
51513.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3B2 | |
Western Blot | |
ETS translocation variant 7, ets variant 7, ets variant gene 7 (TEL2 oncogene), TEL-2, TEL2 oncogene, TEL2ETS-related protein Tel2, TELBEts transcription factor TEL-2b, Tel-related Ets factor, transcription factor ets, transcription factor ETV7, Transcription factor Tel-2, TREF | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction