Learn More
ETV7 Mouse anti-Human, Clone: OTI3B2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802936

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The ETS (E26 transformation-specific) family of transcription factors, of which ETV7 is a member, is a large group of evolutionarily conserved transcriptional regulators that play an important role in a variety of cellular processes throughout development and differentiation, and are involved in oncogenesis as well.Specifications
ETV7 | |
Monoclonal | |
Unconjugated | |
ETV7 | |
TEL-2, TEL2, TELB | |
Mouse | |
Affinity Chromatography | |
RUO | |
51513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y603 | |
ETV7 | |
Human recombit protein fragment corresponding to amino acids 90-341 of human ETV7 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.