Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETV7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592853 Supplier Invitrogen™ Supplier No. PA592853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQEGELAISP ISPVAAMPPL GTHVQARCEA QINLLGEGGI CKLPGRLRIQ PALWSREDVL HWLRWAEQEY SLPCTAEHGF EMNGRALCIL TKDDFRHRAP SSGDVLYELL QYIKTQRRAL VCGPFFGGIF RLKTPTQHSP VPPEEVTGPS QMDTRRGHLL QPPDPGLTSN FGHLDDPGLA RWTPGKEESL NLCHCAELGC RTQGVCSFPA; Positive Samples: U-937, A-431, U-251MG, HeLa; Cellular Location: Nucleus.

The ETS (E26 transformation-specific) family of transcription factors, of which ETV7 is a member, is a large group of evolutionarily conserved transcriptional regulators that play an important role in a variety of cellular processes throughout development and differentiation, and are involved in oncogenesis as well (Potter et al., 2000 [PubMed 10828014]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETV7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ETV7
Gene Accession No. Q9Y603
Gene Alias Ets transcription factor TEL-2b; ETS translocation variant 7; ETS variant 7; ets variant gene 7 (TEL2 oncogene); ETS-related protein Tel2; ETV7; TEL2; TEL-2; TEL2 oncogene; TELB; tel-related Ets factor; Transcription factor ETV7; Transcription factor Tel-2; TREF
Gene Symbols ETV7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human ETV7 (NP_0572191).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51513
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.