Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ETV7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530624
Description
Recommended positive controls: U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The ETS (E26 transformation-specific) family of transcription factors, of which ETV7 is a member, is a large group of evolutionarily conserved transcriptional regulators that play an important role in a variety of cellular processes throughout development and differentiation, and are involved in oncogenesis as well (Potter et al., 2000 [PubMed 10828014]).
Specifications
ETV7 | |
Polyclonal | |
Unconjugated | |
ETV7 | |
Ets transcription factor TEL-2b; ETS translocation variant 7; ETS variant 7; ets variant gene 7 (TEL2 oncogene); ETS-related protein Tel2; ETV7; TEL2; TEL-2; TEL2 oncogene; TELB; tel-related Ets factor; Transcription factor ETV7; Transcription factor Tel-2; TREF | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51513 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.62 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y603 | |
ETV7 | |
Recombinant protein encompassing a sequence within the center region of human ETV7. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction