Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eukaryotic translation initiation factor 1 Antibody (OTI2B9), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271554UV
Description
Eukaryotic translation initiation factor 1 Monoclonal specifically detects Eukaryotic translation initiation factor 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Eukaryotic translation initiation factor 1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EIF1 | |
Full length recombinant protein of human Eukaryotic translation initiation factor 1 (NP_005792) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2B9 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
10209.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction