Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eukaryotic translation initiation factor 5B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $485.00
Specifications
Antigen | Eukaryotic translation initiation factor 5B |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Eukaryotic translation initiation factor 5B Polyclonal specifically detects Eukaryotic translation initiation factor 5B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Eukaryotic translation initiation factor 5B | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
9669 | |
IgG | |
Affinity purified |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
DKFZp434I036, eukaryotic translation initiation factor 5B, FLJ10524, IF2Translation initiation factor IF-2, KIAA0741eIF-5B, translation initiation factor IF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 901-1220 of human Eukaryotic translation initiation factor 5B (NP_056988.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title