Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Euramco Ramfan™ EFi150XX Conductive Duct
SDP

Catalog No. 19817744
Change view
Click to view available options
Length (English) Duct:
180 in.
300 in.
Length (Metric) Duct:
457.2 cm
762 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Duct Length (Metric) Duct
19-817-744 300 in. 762 cm
19-822-707 180 in. 457.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-817-744 Supplier Euramco Supplier No. FDT1625CBR

For safe use in adverse atmospheres

  • High-strength
  • Lightweight anti-static glass reinforced ABS housing
  • IP55 rain tested switch enclosure
  • For hazardous or explosive gases safety

Specifications

Color Black
Diameter (English) Fan 16 in.
Diameter (Metric) Fan 40.64 cm
Product Type Anti-static Duct
For Use With (Equipment) Ramfan electric blower
Length (English) Duct 300 in.
Length (Metric) Duct 762 cm
Material Conductive PVC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.