Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(II) fluoride, anhydrous, 99.9% (metals basis)

Catalog No. AA4173702
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4173702 0.5 g
AA4173704 2 g
AA4173709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4173702 Supplier Thermo Scientific Chemicals Supplier No. 04173702
Add to Cart

CAS: 14077-39-5 | EuF2 | 189.961 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14077-39-5
Molecular Formula EuF2
Molecular Weight (g/mol) 189.961
MDL Number MFCD01866333
InChI Key YLDGIMHVWSTRML-UHFFFAOYSA-L
Synonym europium ii fluoride, acmc-20akvn, europium ii fluoride, anhydrous
PubChem CID 21225127
IUPAC Name europium(2+);difluoride
SMILES [F-].[F-].[Eu+2]

Specifications

Boiling Point >2400°C
Melting Point 1380°C
Physical Form Powder
Quantity 0.5 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 189.96
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.495 g/mL
Assay (metals basis)
Chemical Name or Material Europium(II) fluoride, Anhydrous
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.