Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Catalog No. p-7049207
Click to view available options
Quantity:
2 g
5 g
10 g

CAS: 181266-82-0 | C15H21EuO6 | 449.29 g/mol

It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181266-82-0
Molecular Formula C15H21EuO6
Molecular Weight (g/mol) 449.29
MDL Number MFCD00192170
InChI Key YMTKZYSHNNXTIJ-UHFFFAOYSA-N
Synonym europium iii 2,4-pentanedionate hydrate reo
PubChem CID 131675702
SMILES [Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 140°C (decomposition)
Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
IUPAC Name europium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
Formula Weight 449.29 (Anhydrous)
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Europium(III) 2,4-pentanedionate hydrate
EINECSNumber 238-185-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.