Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) fluoride, 99.99%, (trace metal basis), Thermo Scientific™

Catalog No. AC316170010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle

 

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-25-8
Molecular Formula EuF3
Molecular Weight (g/mol) 208.96
MDL Number MFCD00010998
InChI Key HPNURIVGONRLQI-UHFFFAOYSA-K
Synonym europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
IUPAC Name europium(3+) trifluoride
SMILES [F-].[F-].[F-].[Eu+3]

Specifications

Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99%
Linear Formula EuF3
Formula Weight 208.96
Grade Trace Metal Basis
Packaging Glass Bottle
Chemical Name or Material Europium(III) fluoride, 99.99%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.