Learn More
Europium(III) oxide, REacton™, 99.99% (REO)
Description
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2291°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Insoluble in water. |
IUPAC Name | dieuropium(3+) trioxidandiide |
Formula Weight | 351.92 |
Sensitivity | Hygroscopic |
Appearance | White powder or chunks |
Show More |
Safety and Handling
EINECSNumber | 215-165-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.