Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) phosphate hydrate, 99.9% (REO)

Catalog No. aa4198903
Click to view available options
Quantity:
1 g
5 g

CAS: 14913-20-3 | EuO4P | 246.93 g/mol

Europium finds application in optical coating and thin film application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-20-3
Molecular Formula EuO4P
Molecular Weight (g/mol) 246.93
MDL Number MFCD05664261
InChI Key KJNZTHUWRRLWOA-UHFFFAOYSA-K
Synonym europium iii phosphate hydrate, europium 3+ hydrate phosphate, europium 3+ phosphate-water 1/1/1, europium 3+ ion hydrate phosphate
PubChem CID 16217374
SMILES [Eu+3].[O-]P([O-])([O-])=O

Specifications

Physical Form Chunks and/or Powder
Quantity 1 g
Linear Formula EuPO4·xH2O
Solubility Information Insoluble in water.
IUPAC Name europium(3+) phosphate
Formula Weight 246.93 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) phosphate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-901-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.