Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) sulfate octahydrate, REacton™, 99.99% (REO)

Catalog No. p-7049208
Click to view available options
Quantity:
5 g
25 g

CAS: 10031-55-7 | Eu2H16O20S3 | 736.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-55-7
Molecular Formula Eu2H16O20S3
Molecular Weight (g/mol) 736.216
MDL Number MFCD00149704
InChI Key PGDXJFDXKJLVTJ-UHFFFAOYSA-H
Synonym europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8
PubChem CID 25021676
IUPAC Name europium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]

Specifications

Melting Point 375°C (in -8H2O)
Physical Form Crystalline
Quantity 5 g
Merck Index 14,3904
Formula Weight 736.23 (592.10 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) sulfate octahydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-903-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.