Description
- Highly stable, easy to use conjugates
- Resistant to aggregation
- No sonication, shaking or vortexing required
- No need for blocking solution
- Possibility to work with low conjugate concentrations
- Conjugates can be run on an immunochromatographic assay as well as a microwell-based assay
Kit Contents:
- 1 Midi vial of 200 nmEuropium - Streptavidin conjugate
Specifications
Specifications
| Quantity | 400 μL |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |