Learn More
Novus Biologicals™ Europium Streptavidin MINI
SDP
Catalog No. 12200001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
80 μL
Catalog No. Quantity
12200001 80 μL
1 options
Catalog No. 12200001 Supplier Novus Biologicals™ Supplier No. 12200001
Europium (Eu) Streptavidin conjugates are manufactured using covalent attachment of Streptavidin to the specially treated surface of 200nm Eu particles.
  • Highly stable, easy to use conjugates
  • Resistant to aggregation
  • No sonication, shaking or vortexing required
  • No need for blocking solution
  • Possibility to work with low conjugate concentrations
  • Conjugates can be run on an immunochromatographic assay as well as a microwell-based assay
Kit Contents:
  • 1 Mini vial of 200 nmEuropium - Streptavidin conjugate.
Specifications
Quantity 80 μL
Content And Storage Storage of components varies. See protocol for specific instructions.