Learn More
Siemens Healthineers Eurotrol GAS-ISE Metabolites
Catalog No. p-8669000
Click to view available options
Level:
1
2
3
6
This item is not returnable. View return policy
This item is not returnable. View return policy
Siemens Healthineers Eurotrol GAS-ISE Metabolites is an assayed blood gas, electrolyte and metabolite reference material to verify the precision and accuracy of the epoc Blood Analysis System
  • pH, gases, electrolytes and metabolites in one ampule
  • Three (3) levels covering the physiologically relevant range
  • Use only one epoc cartridge for each control check
  • Optimized for epoc with physiological target values
  • Developed to verify calibration, operating temperature and other performance characteristics
  • Now includes BUN and TCO2
TRUSTED_SUSTAINABILITY
Specifications
Level 1
Quantity 10 x 2.5 mL Vials
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.