Learn More
Novus Biologicals™ EV MiniSEC Columns
SDP
Catalog No. NB555993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 Columns
20 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB555993 20 Columns
NB557255 10 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB555993 Supplier Novus Biologicals™ Supplier No. NBP34104320COLUMNS
Only null left
Add to Cart
Add to Cart
size exclusion chromatography (SEC) columns designed to isolate extracellular vesicles (EVs) from small volume samples
Specifications
Content And Storage Store at 4°C.
Quantity 20 Columns