Learn More
Bullard™ EVA PAPR System, Double Bib Hoods For Head Protection
For use with hard hats, EVA system includes PAPR, EVA blower, battery, charger, breathing tube. Choose from Tychem QC or Tychem SL double bib hoods.
$2411.00
Specifications
Battery Life | 7 to 8 hours |
---|---|
Belt Material | Foam |
Product Type | PAPR Hard Hat System |
Includes | PAPR system, EVA blower, battery, charger, breathing tube |
Material (Lens) | Acetate |
Catalog Number | Mfr. No. | Facepiece/Headtop Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Facepiece/Headtop Model | Price | Quantity & Availability | |||||
17-989-613
![]() |
Bullard™
EVA20TICHT |
Tychem QC hood |
Each of 1 for $2,411.00
|
|
|||||
Description
- Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
- Easy-to-read fuel gauge displays remaining battery capacity at a glance
- Battery status can be checked at any time during use
- Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
- Two distinct audible alarms notify wearers of low battery and low air flow
- Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
- Powered by a long-lasting 10,000-hour brushless motor
- Single filter design for easy use
- Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Specifications
7 to 8 hours | |
PAPR Hard Hat System | |
Acetate |
Foam | |
PAPR system, EVA blower, battery, charger, breathing tube |