Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVA1 Mouse anti-Mouse, Human , PE, Clone: G9P3-1, BD Biosciences™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 567106

Catalog No. BDB567106


Only null left
Add to Cart

Description

Description

EVA1 Monoclonal antibody specifically detects EVA1 in Mouse, Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EVA1
Monoclonal
PE
epithelial V-like antigen 1; Eva; Eva1; myelin protein zero-like protein 2; Mpzl2
Mouse EVA1 Recombinant Protein
RUO
Mouse, Human
IgG1 κ
Flow Cytometry
G9P3-1
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
0.1 mg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.