Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVA1/MPZL2 Antibody (OTI2C7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271553APC
Description
EVA1/MPZL2 Monoclonal antibody specifically detects EVA1/MPZL2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
EVA1/MPZL2 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
10205 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry | |
OTI2C7 | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
EVAEpithelial V-like antigen 1EVA1myelin protein zero-like protein 2, myelin protein zero-like 2 | |
Full length human recombinant protein of human MPZL2(NP_658911) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction