Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVA1/MPZL2 Antibody (OTI2C7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271553IR
Description
EVA1/MPZL2 Monoclonal specifically detects EVA1/MPZL2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
EVA1/MPZL2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MPZL2 | |
Full length recombinant protein of human EVA1/MPZL2 (NP_658911) produced in HEK293T cells. | |
0.1 mL | |
10205.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2C7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EVAEpithelial V-like antigen 1EVA1myelin protein zero-like protein 2, myelin protein zero-like 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction