Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EVA1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569180
Description
This target displays homology in the following species: Dog: 89%; Goat: 80%; Guinea Pig: 87%; Horse: 100%; Human: 100%; Mouse: 82%; Rabbit: 86%; Rat: 93%; Yeast: 100%.
EVA1B is a protein coding gene.
Specifications
EVA1B | |
Polyclonal | |
Unconjugated | |
EVA1B | |
2610027C15Rik; C1orf78; eva-1 homolog B; eva-1 homolog B (C. elegans); EVA1B; Fam176b; family with sequence similarity 176, member B; protein eva-1 homolog B; Protein FAM176B; RGD1308876 | |
Rabbit | |
Affinity chromatography | |
RUO | |
362597 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
EVA1B | |
synthetic peptide directed towards the middle region of human EVA1B. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction