Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVC Monoclonal Antibody (3C4)
GREENER_CHOICE

Catalog No. PIMA524405
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524405 Supplier Invitrogen™ Supplier No. MA524405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: VLERQRLMQC DLEEEENVRA TEAVVALCQE LYFSTVDTFQ KFVDALFLQT LPGMTGLPPE ECDYLRQEVQ ENAAWQLGKS NRFRRQQWKL FQELLEQDQQ VWMEECALSS.

This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C4
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Evc
Gene Accession No. P57679
Gene Alias DWF-1; Ellis van Creveld gene homolog; Ellis van Creveld gene syndrome; Ellis van Creveld protein; Ellis van Creveld syndrome; ellis-van Creveld syndrome protein; Ellis-van Creveld syndrome protein homolog; EVC; EvC ciliary complex subunit 1; EvC protein (Ellis-van Creveld syndrome); EVC1; EVCL
Gene Symbols Evc
Host Species Mouse
Immunogen EVC (NP_055371, 493 a.a. ~ 602 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2121
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.