Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553412 Supplier Invitrogen™ Supplier No. PA553412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETGSPSRRRK REVQMSKDKE AVDECEPPSN SNITAFALKA KVIYPINQKF RPLADGSSNP SLHENLKQAV LPHQPVEASP SSSLGSLSQG EKDDCSSSSS VHS Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 66%.

This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVC
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Evc
Gene Accession No. P57679
Gene Alias DWF-1; Ellis van Creveld gene homolog; Ellis van Creveld gene syndrome; Ellis van Creveld protein; Ellis van Creveld syndrome; ellis-van Creveld syndrome protein; Ellis-van Creveld syndrome protein homolog; EVC; EvC ciliary complex subunit 1; EvC protein (Ellis-van Creveld syndrome); EVC1; EVCL
Gene Symbols Evc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EVC. Recombinant protein control fragment (Product #RP-89973).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2121
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.