Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561656 Supplier Invitrogen™ Supplier No. PA561656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYESKLEPLP FTSADGVNED LSLNDQMIDI LSSEDPGSML QALEELEIAT LNRADADLEA CRTQISKDII ALLLKNLTSS GHLSPQV Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.

This gene encodes a protein that functions in bone formation and skeletal development. Mutations in this gene, as well as in a neighboring gene that lies in a head-to-head configuration, cause Ellis-van Creveld syndrome, an autosomal recessive skeletal dysplasia that is also known as chondroectodermal dysplasia. Mutations in this gene also cause acrofacial dysostosis Weyers type, also referred to as Curry-Hall syndrome, a disease that combines limb and facial abnormalities. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EVC2
Gene Accession No. Q86UK5
Gene Alias Ellis van Creveld syndrome 2; ellis-van Creveld syndrome protein 2; EvC ciliary complex subunit 2; EVC2; LBN; limbin; WAD
Gene Symbols EVC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EVC2. Recombinant protein control fragment (Product #RP-99913).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.