Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVC2 Polyclonal Antibody

EVC2 Polyclonal Antibody
Supplier: Thermo Scientific PA550658
Description
EVC2 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
EVC2 | |
Polyclonal | |
Unconjugated | |
EVC2 | |
Ellis van Creveld syndrome 2; ellis-van Creveld syndrome protein 2; EvC ciliary complex subunit 2; EVC2; LBN; limbin; WAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132884 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q86UK5 | |
EVC2 | |
Synthetic peptide corresponding to a region derived from internal residues of human Ellis van Creveld syndrome 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction