Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eve Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569510
Description
This target displays homology in the following species: Fruit fly: 100%.
eve gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
eve | |
Polyclonal | |
Unconjugated | |
eve | |
10.5; 10.9; 14.10; 20.35; CG2328; CG2328-PA; complementation group F; Dmel\CG2328; Dmel_CG2328; dm-eve; E(eve); Eve; eve PRE/TRE element; eve2; even; even skipped; evenskip; even-skiped; evenskipped; even-skipped; eve-PA; F; group V; group VI; l(2)46Ce; l(2)46CFg; l(2)46CFh; l(2)46CFj; l(2)46CFp; l(2)46Cg; lethal(2)46Ce; Segmentation protein even-skipped; V; VI | |
Rabbit | |
Affinity chromatography | |
RUO | |
36039 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P06602 | |
eve | |
synthetic peptide corresponding to a region of fruit fly eve (aa 1-50). | |
100 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction