Learn More
Caplugs™ (Evergreen Scientific) Empty Histoware™ Vials
Catalog No. p-3238159
Click to view available options
Capacity (Metric):
60 mL
90 mL
Unfilled shatterproof vials for histology work
  • Formulated polypropylene provides unmatched vial clarity and tensile strength
  • Shatterproof if accidentally dropped
  • Green polyethylene screwcaps have molded-in sealing rings for virtually leakproof seals
Specifications
Capacity (Metric) 60 mL
Closure Color Green
Closure Material Polyethylene
Closure Type Screw Cap
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.