Learn More
Caplugs™ (Evergreen Scientific) Microcentrifuge Tubes
Catalog No. 055559 Shop All Caplugs Products
Change view
Click to view available options
Closure Type:
Not Included
Snap Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
05-555-9 Snap Cap
05-558-33A Not Included
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-555-9 Supplier Caplugs™ (Evergreen Scientific) Supplier No. 2143410080
Add to Cart
Add to Cart
Fit all standard microcentrifuges.
  • Precision molded from premium grade polypropylene
  • 1.5mL capacity
  • Lot tested for dimensional uniformity, tensile strength, and physical appearance
Specifications
Capacity (Metric) 1.5 mL
Material Polypropylene
Closure Type Snap Cap
Max. RCF 12,000 x g