Learn More
Caplugs™ (Evergreen Scientific) Sputocol™ Sputum Collection System
Catalog No. 055540 Shop All Caplugs Products
Click to view available options
Includes:
Conical tubes
Freestanding tubes
This item is not returnable. View return policy
This item is not returnable. View return policy
Polypropylene
  • Individually packed
  • Withstand 6000 × g
  • Graduated
  • With patient label
Specifications
Product Type Sputum Collection System
Graduated Yes
Includes Freestanding tubes
Material Polypropylene
Max. RCF 6000 x G
Volume (Metric) 50 mL
Volume (English) 1.69 oz.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.