Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EVG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | EVG1 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Target Species | Human |
Description
EVG1 Polyclonal specifically detects EVG1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
EVG1 | |
Unconjugated | |
Human | |
chromosome 22 open reading frame 23, dJ1039K5.6, FLJ32787 | |
C22orf23 | |
IgG | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
84645 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LQCSPTSSQRVLPSKQIASPIYLPPILAARPHLRPANMCQANGAYSREQFKPQATRDLEKEKQRLQNIFATGKDMEERKRKAPPARQKAPAPELDRFEELVKEIQERKEFLADMEALGQGKQYRGIILAEISQKLREMEDIDHRRSEE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title