Learn More
Description
Specifications
Specifications
| Antigen | EVI-1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Peptide ELISA 1:2000 |
| Formulation | Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide |
| Gene Accession No. | NP_001098547.3 |
| Gene Alias | AML1-EVI-1, AML1-EVI-1 fusion protein, ecotropic viral integration site 1, Ecotropic virus integration site 1 protein homolog, EVI-1, EVI1MDS1-EVI1, MDS1, MDS1 and EVI1 complex locus, MDS1 and EVI1 complex locus protein EVI1, MDS1 and EVI1 complex locus protein MDS1, MGC163392, MGC97004, myelodysplasia syndrome 1, Myelodysplasia syndrome 1 protein, Myelodysplasia syndrome-associated protein 1, oncogene EVI1, PRDM3, zinc finger protein Evi1 |
| Gene Symbols | MECOM |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
