Learn More
Invitrogen™ EVI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596384
Description
Positive Samples: Mouse kidney, Mouse heart; Cellular Location: Nucleus, Nucleus speckle Immunogen sequence: MRSKGRARKL ATNNECVYGN YPEIPLEEMP DADGVASTPS LNIQEPCSPA TSSEAFTPKE GSPYKAPIYI PDDIPIPAEF ELRESNMPGA GLGIWTKRKI EVGEKFGPYV GEQRSNLKDP SYGWEILDEF YNVKFCIDAS QPDVGSWLKY IRFAGCYDQH NLVACQINDQ IFYRVVADIA PGEELLLFMK.
EVI1 (MECOM) is a transcriptional regulator and oncoprotein that may be involved in hematopoiesis, apoptosis, development, cell differentiation, and proliferation. It can interact with CTBP1, SMAD3, CREBBP, KAT2B, MAPK8, and MAPK9. The EVI1 gene can undergo translocation with the AML1 gene, resulting in overexpression of the EVI1 gene and the onset of leukemia. Several transcript variants encoding a few different isoforms have been found for this gene.
Specifications
EVI1 | |
Polyclonal | |
Unconjugated | |
MECOM | |
AML1-EVI-1; AML1-EVI-1 fusion protein; D630039M04Rik; ecotropic viral integration site 1; ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI1; Evi-1; Histone-lysine N-methyltransferase MECOM; Jbo; Mds; MDS1; MDS1 and EVI1 complex locus; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; Mds1-Evi1; Mecom; MGC163392; MGC97004; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; myelodysplasia syndrome-associated protein 1; oncogene EVI1; OTTHUMP00000213163; OTTHUMP00000213164; OTTHUMP00000213165; OTTHUMP00000213187; OTTHUMP00000213192; PR domain 3; PRDM3; RUSAT2; zinc finger protein Evi1; Znfpr1b1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2122, 294924 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03112 | |
MECOM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human MECOM (NP_0049822). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.