Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115561 Supplier Invitrogen™ Supplier No. PA5115561
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MDS1.

EVI1 (MECOM) is a transcriptional regulator and oncoprotein that may be involved in hematopoiesis, apoptosis, development, cell differentiation, and proliferation. It can interact with CTBP1, SMAD3, CREBBP, KAT2B, MAPK8, and MAPK9. The EVI1 gene can undergo translocation with the AML1 gene, resulting in overexpression of the EVI1 gene and the onset of leukemia. Several transcript variants encoding a few different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MECOM
Gene Accession No. P14404, Q03112
Gene Alias AML1-EVI-1; AML1-EVI-1 fusion protein; D630039M04Rik; ecotropic viral integration site 1; ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI1; Evi-1; Histone-lysine N-methyltransferase MECOM; Jbo; Mds; MDS1; MDS1 and EVI1 complex locus; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; Mds1-Evi1; Mecom; MGC163392; MGC97004; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; myelodysplasia syndrome-associated protein 1; oncogene EVI1; OTTHUMP00000213163; OTTHUMP00000213164; OTTHUMP00000213165; OTTHUMP00000213187; OTTHUMP00000213192; PR domain 3; PRDM3; RUSAT2; zinc finger protein Evi1; Znfpr1b1
Gene Symbols MECOM
Host Species Rabbit
Immunogen A synthesized peptide derived from human MECOM(Accession Q03112), corresponding to amino acid residues A1013-N1063.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14013, 2122, 294924
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.