Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI1 Polyclonal Antibody, Huabio

Catalog No. 502544591
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4591 Supplier HUABIO RESEARCH INC Supplier No. HA500173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EVI1 Polyclonal antibody specifically detects EVI1 in Human samples. It is validated for Western Blot

Specifications

Antigen EVI1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene MECOM
Gene Accession No. Q03112
Gene Alias AML1-EVI-1; AML1-EVI-1 fusion protein; D630039M04Rik; ecotropic viral integration site 1; ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI1; Evi-1; Histone-lysine N-methyltransferase MECOM; Jbo; Mds; MDS1; MDS1 and EVI1 complex locus; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; Mds1-Evi1; Mecom; MGC163392; MGC97004; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; myelodysplasia syndrome-associated protein 1; oncogene EVI1; OTTHUMP00000213163; OTTHUMP00000213164; OTTHUMP00000213165; OTTHUMP00000213187; OTTHUMP00000213192; PR domain 3; PRDM3; RUSAT2; zinc finger protein Evi1; Znfpr1b1
Host Species Rabbit
Immunogen Recombinant protein within human Evi-1 aa 1-200.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2122
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.